Synthware Condenser, Air Reflux, Threaded, Microscale, Kemtech America
60004-116
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- ST Joint:14/10
- Cat. no.:60004-116
- Effective Length:100 mm
- Supplier No.:C221410
Air reflux condenser for microscale applications.
- Heavy wall borosilicate construction
- Threaded upper joint
- Open cap included
Supplied with a nylon compression cap and Viton® O-ring.