Synthware Sublimator, 25 ml, with 2 mm Glass Stopcock, Kemtech America
Catalog # 60004-098
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Sublimator
- Cat. no.:60004-098
- Supplier No.:D232440G
Specifications
About this item
A sublimator with a 2mm glass stopcock and a 25mL long-neck flask for small volume efficiency.
- Heavy wall borosilicate construction
- Long neck flask
- Clip included
- All glass stopcock