Accessories for Spherical Pinch Clamps, Kemtech America
60004-974
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Description:Metal Clamp for 18/8 Spherical Joint
- Cat. no.:60004-974
- Supplier No.:CC0018
Stainless stel pinch clamp for spherical joints.
- Stainless Steel
- Tensioner Knob
For use with 18/8 ball joint or #9 O-Ring joint.