ContactUsLinkComponent
Compresstome® Legacy Model Blade Holder
Catalog # 77435-418
undefined
Compresstome® Legacy Model Blade Holder
Catalog # 77435-418
Supplier Number:  VFBHVMLEGACYNC
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Compresstome legacy model blade holder
  • Cat. no.:
    77435-418
Specifications
About this item
Replacement blade holder for the Compresstome vibrating microtome.
  • Compatible with models VF-200, VF-200-0Z, VF-300, and VF-300-0Z
A replacement blade holder for legacy model Comprestomes. This device is compatible with Compresstome models VF-200, VF-200-0Z, VF-300, and VF-300-0Z. Includes one blade holder, one small Allen wrench, and one small screw.