ContactUsLinkComponent
EnSURE™ Touch Monitoring System, EnSURE™ Touch Monitoring System
EnSURE™ Touch Monitoring System, EnSURE™ Touch Monitoring System
Catalog # CA76422-850
undefined
EnSURE™ Touch Monitoring System, EnSURE™ Touch Monitoring System
Catalog # CA76422-850
Supplier Number:  ETOUCH
Specifications
  • Description:
    EnSURE™ touch monitoring system
  • Cat. no.:
    CA76422-850
Specifications
About this item
EnSURE® Touch is an advanced monitoring system that collects, analyzes, and reports data from a variety of long-trusted quality tests, providing rapid and accurate sanitation verification data. Featuring a 5" touchscreen, wireless sync technology, and cloud-based software. The EnSURE® Touch is designed to adapt to your workplace, providing the data you need for complex multi-location supply chain monitoring, audit and risk management, and recall prevention.
  • Verify sanitation of surfaces, liquids, and product testing in as little as ten seconds
  • Test and compare the effectiveness of sanitation products and procedures
  • Train and monitor sanitation crew and QC/QA employees
  • Support SSOP and objectively prove cleaning standards have been met
  • Quickly identify when replaceable parts like belts and brushes should be replaced
  • Identify problem areas in your facility that need more attention
  • Provide on-the-spot training and feedback
The EnSURE® Touch monitoring system is designed to be used for multiple quality and food safety tests. Users can use the system for ATP hygiene monitoring or have an expanded program that incorporates rapid microorganism tests for environmental monitoring and finished product testing. Additional tests for pasteurization and cooking proficiency are also available.
EnSURE® Touch makes no compromises when it comes to durability. The rugged outer shell and shatter-proof touchscreen keep EnSURE® Touch safe in the harshest environments. For extra protection, every system comes with a complimentary carrying case with a shoulder strap for hands-free transportation to test locations in your facility.
Industries: Breweries, wineries and distilleries, Cannabis QA testing, cosmetics, environmental, food safety.