Acetone for HPLC
Glass bottle, 4 L
Catalog # RCRSOA0011-4C
Supplier: Ricca Chemical
Supplier #: RSOA0011-4C
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Storage Temperature:Ambient
- UN:1090
- ADR:3,II
Specifications
Specification Test Results
- Assay:min 99.5 %
- Color (APHA):max 10
- Solubility in Water:To Pass Test
- Isopropyl Alcohol:max 0.05 %
- Methanol (Methyl Alcohol):max 0.05 %
- Water (H₂O):max 0.5 %
- Aldehyde (as HCHO):max 0.002 %
- Residue after evaporation (Non-Volatile Matter):max 0.001 %
- Substances Reducing Permanganate:To Pass Test
- Titratable Acid:max 0.0003 meq/g
- Titratable Base:max 0.0006 meq/g
- Absorbance at 330 nm:0.00-1.00
- Absorbance at 340 nm:0.000-0.100
- Absorbance at 350 nm:0.000-0.020
- Absorbance at 400 nm:0.000-0.010
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:RCRSOA0011-4C
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



