Order Entry
Canada
ContactUsLinkComponent
Syringe for Automatic Liquid Handling, AD
Syringe for Automatic Liquid Handling, AD
Catalog # 77506-152
Syringe for Automatic Liquid Handling, AD
Catalog # 77506-152
Supplier Number:  2605813

Specifications

  • Volume:
    250 µl
  • Description:
    Plunger tip: PTFE seal
  • Cat. no.:
    77506-152

Specifications

About this item

Syringes for automatic liquid handling made up of borosilicate 3.3.

  • 60 mm stroke

Precision: < ±1% of the volume
Front fitting: M8×0.75