Order Entry
Canada
ContactUsLinkComponent
Synthware Double Bank Manifold with #15 O-Ring Joints, Kemtech America
Synthware Double Bank Manifold with #15 O-Ring Joints, Kemtech America
Catalog # 60004-596
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Double Bank Manifold with #15 O-Ring Joints, Kemtech America
Catalog # 60004-596
Supplier:  Kemtech America
Supplier Number:  M330804
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Double bank vacuum manifold
  • Environmentally Preferable:
  • Cat. no.:
    60004-596
  • No. of Ports:
    4
  • Supplier No.:
    M330804

Specifications

About this item

Forward facing 0 to 8 mm high vacuum valves allow for easy operation within a fume hood.

  • Hi Vacuum Valves
  • Heavy wall borosilicate construction
  • 4 ports
  • #15 O-ring joints
  • Double bank

Ports are spaced 100 mm center to center. Accessory items must be ordered separately.