ContactUsLinkComponent
Syringes for HPLC Autosamplers
Syringes for HPLC Autosamplers
Catalog # 77506-150
Syringes for HPLC Autosamplers
Catalog # 77506-150
Supplier Number:  2601702
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Ext.Ø:
    0.72 mm
  • Capacity:
    100 µl
  • Environmentally Preferable:
  • Cat. no.:
    77506-150
  • Gauge:
    G22S
Specifications
About this item
  • HPLC valves
  • Autosamplers
  • Dosage of aggressive fluids
  • Glass barrel: Borosilicate 3.3
  • Plunger tip: PTFE
Plunger: Stainless steel with PTFE-seal
Glass barrel: Borosilicate 3.3