SGE CYDEX-B GC Capillary Columns
Supplier: Trajan Scientific and Medical
Certificates
About this item
The SGE CYDEX-B GC Capillary Column provides a great low-polarity option for GC analyses. Recommended for: Separation of enantiomeric compounds found in natural products.
- Permethylated Beta-Cyclodextrin phase for the separation of chiral compounds
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.
Specifications
- Catalog No:
- 60361-944
- 60361-628
- 60361-630
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Temperature limit:
- 240 °C
- 240 °C
- 240 °C