SGE BPX1 GC Capillary Columns
Supplier: Trajan Scientific and Medical
Certificates
About this item
The SGE BPX1 GC capillary column provides a great non-polar option for GC analyzes. Recommended for: ASTM methods D2887 and D6532.
- Non-polar column
- Dimensionally stabilized phase
- Low bleed
- Specifically designed for high temperature hydrocarbon analysis
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.
Specifications
- Catalog No:
- 14233-472
- 60361-648
- 14226-054
- 14226-030
- Description:
- 14233-472
- 60361-648
- 14226-054
- 14226-030
- Polarity:
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Temperature limit:
- 400 °C
- 370 °C
- 370 °C
- 400 °C