ContactUsLinkComponent
VWR® Lab Pen black ink
VWR® Lab Pen black ink
Catalog # 77507-062
Supplier:  VWR International
VWR® Lab Pen black ink
Catalog # 77507-062
Supplier:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Black
  • Description:
    VWR Lab Pen, 12/box
  • Environmentally Preferable:
  • Cat. no.:
    77507-062
Specifications
About this item
VWR®, part of Avantor has industry leading Anatomical Pathology consumables available your lab.
  • Designed exclusively for lab techs, this original formulation does not dry out, and withstands exposure to common processing chemicals.
  • Excellent for both slides and embedding cassettes.