WHEATON® Serum Vials, Borosilicate Glass, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Tubular design provides excellent clarity and dimensional consistency from vial to vial.
- Autoclavable
Clear vials manufactured from low extractable borosilicate glass.Amber glass protect light-sensitive samples.
Specially designed bottom radius adds strength for lyophilization applications.
Certifications: Amber glass conforms to USP Type I requirements.Borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Packaging: Vials are packaged in shrink-wrapped modules reduce particulate contamination.
Related Information
Certificates
Search by Lot Number16171-291_1631534
Conformity declaration
Wheaton Vial GUide
Product Literature
Customers Who Bought This Also Bought