Rtx®-1701 GC Columns, Restek
Supplier: Restek
Certificates
About this item
For the analysis of pesticides, PCBs, solvents, and drugs.
- Internal dimensions and film thickness vary between models
- Low- to mid-polarity column
- Different elution order from Rtx®-1 or Rtx®-5 columns due to higher polarity
A low- to mid-polarity column with a different elution order than less polar Rtx®-1 or Rtx®-5 columns. Not recommended for strongly acidic compounds since cyano phases may show non-linearity at ppm levels. Phase composition: crossbonded 14% cyanopropylphenyl, 86% dimethylpolysiloxane. Recommended for EPA method 608.
Specifications
- Catalog No:
- RK12010
- 10850-246
- 10850-248
- 10850-250
- RK12073
- RK12055
- 10803-424
- RK12058
- RK12057
- RK12020
- RK12025
- RK12026
- 10059-544
- RK12011
- RK42012
- RK12059
- RK12088
- RK12024
- RK12041
- RK12027
- RK12035
- 75851-742
- RK12008
- RK12085
- RK12009
- RK12072
- RK12069
- RK12070
- RK12021
- RK12030
- RK12082
- RK12023
- RK12038
- RK12037
- RK12040
- RK12039
- RK12042
- RK12050
- 10850-848
- RK12052
- RK12051
- 10849-562
- RK12054
- RK12053
- 10060-012
- RK12066
- RK12056
- Description:
- RK12010
- RK12073
- RK12055
- 10803-424
- RK12058
- RK12057
- RK12020
- RK12025
- RK12026
- 10059-544
- RK12011
- RK12059
- RK12088
- RK12024
- RK12041
- RK12027
- RK12035
- RK12008
- RK12085
- RK12009
- RK12072
- RK12069
- RK12070
- RK12021
- RK12030
- RK12082
- RK12023
- RK12038
- RK12037
- RK12040
- RK12039
- RK12042
- RK12050
- RK12052
- RK12051
- RK12054
- RK12053
- RK12066
- RK12056
- Temperature limit:
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+260 °C
- –20...+260 °C
- –20...+280 °C
- –20...+270 °C
- –20...+280 °C
- –20...+280 °C
- –60...+320 °C
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+250 °C
- –60...+320 °C
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+250 °C
- –20...+280 °C
- –20...+250 °C
- –20...+280 °C
- –20...+260 °C
- –20...+260 °C
- –20...+260 °C
- –20...+280 °C
- –20...+280 °C
- –20...+250 °C
- –60...+320 °C
- –20...+280 °C
- –20...+270 °C
- –60...+270 °C
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+280 °C
- –20...+270 °C
- –20...+280 °C
- –20...+280 °C
- –20...+260 °C
- –20...+280 °C
- –20...+280 °C
- –20...+260 °C
- –20...+260 °C