ContactUsLinkComponent
UV Pocket Microscope Kit with 24 Prepared Slides
  470355-714
UV Pocket Microscope Kit with 24 Prepared Slides
  470355-714
 :  MP-250BUN
  • Magnification:
    100 - 120×
  • Model:
    MP-250BUN
  • Description:
    Carson MicroFlip™ LED and UV pocket microscope kit
  • Cat. no.:
    470355-714
  • Includes:
    24 prepared slides, flip down slide base, smartphone digiscoping clip
The MicroFlip™ 100 to 250× LED and UV lighted pocket microscope with flip down side base and smartphone digiscoping clip MP-250 is an extremely powerful LED lit pocket microscope that contains a unique flip-down slide base with an aspheric lens system for ultra high definition image quality. The high-power magnification ranges from 100x-250x power. The MicroFlip™ also contains a built-in UV flashlight, making for the perfect STEM kit for scientific research, biology/microbiology, jewelry and so much more.
  • Powered by 1 AA battery (not included)
  • Cost effective
  • Educational field microscope for multiple hobbies, science experiments, learning, and exploration
The MicroFlip™ comes included with a smartphone digiscoping clip, 12 unique insect specimen prepared slides, 12 unique animal specimen prepared slides, and a wrist strap for convenient portability on all your field studies.
Use the smartphone digiscoping clip to take pictures and record video of your subject matter using the built-in camera application on your smartphone. The smartphone digiscoping clip will work with all smartphones. Thicker phone cases may need to be removed before attaching the digiscoping clip.