Orders LinkContactUsLinkComponent
Electro-Gel™ Electrolyte Replenishers
Electro-Gel™ Electrolyte Replenishers
Supplier:   Bio-Serv
Electro-Gel™ Electrolyte Replenishers
Supplier:  Bio-Serv
Certificates
Electro-Gel™ Electrolyte Replenishers
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Gelled, oral electrolyte hydration supplement. Provides balanced electrolyte and glucose levels to provide energy, nutrients and hydration to accelerate recovery.
  • Economical and easy to use requiring no refrigeration or special storage
  • Highly palatable with a sweet, salty orange or strawberry flavor
  • Comparable to Pedialyte and PRANG in gel form
  • Suitable for a variety of research species
  • Thermoreversible
  • Gamma Irradiated
Shelf life of 12 months, cool dry storage.
Ingredients: Purified water, dextrose, gelatin, L-ascorbic acid phosphate, sodium chloride, sodium bicarbonate, citric acid, potassium chloride, flavoring, natural color, potassium sorbate, acesulfame potassium.
Product Family Options
Product Information
  • Size
    Packaging
    Availability
    Price
  • 1 oz.
    16 cups/pack
    Log in for availability
    Log in for availability
    Pack of 16
    Pack of 16 - $70.72
    $70.72
    Pack of 16
       $70.72
    Supplier Number: EGS16P-1
  • 1 oz.
    60 cups/case
    Discontinued
    Supplier Number: EGS-1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 1 oz.
    60 cups/case
    Log in for availability
    Log in for availability
    Case of 60 (1/Itm/Bx)
    Case of 60 (1/Itm/Bx) - $220.80
    $220.80
    Case of 60 (1/Itm/Bx)
       $220.80
    Minimum order quantity is 1
    Supplier Number: EGS-1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.