ContactUsLinkComponent
SP Bel-Art Scintillation Vial Rack, Polypropylene, 24-Position, Bel-Art Products, a part of SP
SP Bel-Art Scintillation Vial Rack, Polypropylene, 24-Position, Bel-Art Products, a part of SP
  66022-503
undefined
SP Bel-Art Scintillation Vial Rack, Polypropylene, 24-Position, Bel-Art Products, a part of SP
  66022-503
 :  F18512-0000
  • Cat. no.:
    66022-503
Rugged, stable polypropylene rack holds 24 scintillation vials (30 mm) for storage or preparation of cocktails.
  • Rugged and stable
  • Polypropylene construction
  • Autoclavable
Scintillation vial rack is resistant to corrosion and convenient for storage, handling, and sample preparation.
Shelf height: 4.4 cm (13/4")