SP Bel-Art Dry-Keeper™ Small, Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Catalog # 74950-336
Supplier: Bel-Art Products, a Part of SP
Specifications
- Volume:0.14 cu. ft. (internal)
- Cat. no.:74950-336
- L×D×H:23 x 20 x 17 cm (87/8 x 77/8 x 65/8")
- Supplier No.:420530000
Specifications
About this item
Small, clear polystyrene desiccator cabinet safely stacks up to 5 units high for maximum use of dry storage space.
- Polystyrene construction
- Door disappears beneath cabinet when open
- Non-electric
- Stackable design
Pull-down door slides out of the way into a compartment located underneath the cabinet.
Made of high-impact polystyrene. Completely transparent for full visibility from any side. Foam rubber gasket forms seal when door is latched with two side roller latches. When opened, door disappears underneath cabinet. May be stacked five high to save bench space.
Ordering information: Ready to use jar of silica gel is included.