SP Bel-Art Dry-Keeper™ Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Perforated shelves are movable to suit your dry storage needs in this desiccator cabinet that stacks up to 5 units high.
- Polystyrene construction
- Adjustable, perforated shelves
- Door disappears beneath cabinet when open
This smoke-tinted polystyrene cabinet has two removable shelves that can be set at four heights.
The pull-down door slides out of the way into a compartment located underneath the cabinet.
Ordering information: Includes a small desiccant tray.
Specifications
- Catalog No:
- 24983-331
- Exterior dimensions:
- 25W×25D×25H cm (927/32×927/32×927/32")