ContactUsLinkComponent
ONPG Disks
ONPG Disks
Catalog # CA77497-262
CAS Number:  
undefined
ONPG Disks
Catalog # CA77497-262
Supplier Number:  Z7321
CAS Number:  
Specifications
  • Description:
    ONGP Disks detect galactosidase
  • Size:
    50 disks/cartridge
  • Cat. no.:
    CA77497-262
Specifications
About this item
ONPG differentiation disk is used to differentiate members of the enterobacteriaceae and other microorganisms based on beta-galactosidase activity.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.