Orders LinkContactUsLinkComponent
Nylon Bones, Petite, Certified
 :   Bio-Serv
Nylon Bones, Petite, Certified
 :  Bio-Serv
Nylon Bones, Petite, Certified
These nylon bones are made of pure virgin nylon.
  • Uniform color and consistency
  • Autoclavable
  • Cagewasher safe
The Half Bone is very economical. Provides gnawing enrichment for rodents. Chocolate brown color provides better visibility in bedding.
 : Certified (Contaminant screened).
Product Family Options
  • Description
    Color
    Length
    Width
    Packaging
    Material
  • Half bone, petite
    Brown
    4.44 cm (1.75")
    2.54 cm (1")
    100/Box
    Nylon
     : K3594
  • Half bone, petite
    Brown
    4.44 cm (1.75")
    2.54 cm (1")
    100/Box
    Nylon
     : K3594
  • Whole bone, petite
    Brown
    10.1 cm (4")
    2.54 cm (1")
    50/Box
    Nylon
     : K3593
  • Whole bone, petite
    Brown
    10.1 cm (4")
    2.54 cm (1")
    50/Box
    Nylon
    Box of 50
    Box of 50 - $135.76
    $135.76
    Box of 50
       $135.76
     : K3593