Synthware Filtration Apparatus with 47 mm Fritted Disc, [ST]40/35 Joint Connections, Kemtech America
Catalog # 60002-266
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Complete Support Assembly
- Cat. no.:60002-266
- Supplier No.:F10401L
Specifications
About this item
Uses 47 mm membranes for filtering corrosive solutions, particulate removal from HPLC solvents, and filtration analysis with solid phase extraction disks.
- Complete Kit
- Heavy wall Borosilicate construction
- Ground glass joint
- Fritted glass filter
Support base has a sealed-in, coarse porosity fritted disc.
Delivery: Complete assembly includes a 300 ml graduated funnel, aluminum clamp, support base, and 1 L filtering flask.