Order Entry
Canada
ContactUsLinkComponent
 
Synthware Distillation Head, Microscale, Kemtech America
Catalog # 60002-160
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Distillation Head, Microscale, Kemtech America
Catalog # 60002-160
Supplier:  Kemtech America
Supplier Number:  D112440G
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Microscale distillation head
  • Cat. no.:
    60002-160
  • Supplier No.:
    D112440G

Specifications

About this item

With 2 mm glass stopcock and [ST] 24/40 joint.

  • Heavy wall borosilicate construction
  • Long neck flask
  • All glass stopcock

Supplied with a special 25 ml long neck flask for small volume efficiency. [ST] joints size: 24/40. Hose ext. Ø: 8 mm.

Ordering information: For replacement flask, contact your Avantor representative.