Order Entry
Canada
ContactUsLinkComponent
Synthware Bubblers with Bottom Hose Connection, Kemtech America
Synthware Bubblers with Bottom Hose Connection, Kemtech America
Catalog # 60000-726
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Bubblers with Bottom Hose Connection, Kemtech America
Catalog # 60000-726
Supplier:  Kemtech America
Supplier Number:  B262910
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Cat. no.:
    60000-726
  • Top ST Joint Size:
    [ST] 29/42
  • Supplier No.:
    B262910

Specifications

About this item

With small ball joints to prevent mineral oil from being sucked back under sudden negative pressure in the system.

  • Heavy wall borosilicate construction
  • Ground glass joint
  • Anti backflow design

Bottom hose connection diameter: 10 mm. Length: 157 mm.