ContactUsLinkComponent
LJ (Lowenstein Jensen) Deep for Catalase Test
Catalog # CA77497-254
CAS Number:  
undefined
LJ (Lowenstein Jensen) Deep for Catalase Test
Catalog # CA77497-254
Supplier Number:  C27
CAS Number:  
Specifications
  • Pack type:
    PC tube
  • Description:
    Deep fill, 20×150 mm, 20 pack
  • Cat. no.:
    CA77497-254
  • Volume:
    5 ml
Specifications
About this item
Egg based media for use in the cultivation and isolation of Mycobacterium spp.