VWR® Talon® Nester Extension Clamp
Catalog # 21572-002
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Description:VWR® Talon® Nester Extension Clamp
- Cat. no.:21572-002
Specifications
About this item
Securely holds 50–102 mm (2–4") diameter large or small glass distillation columns and odd-shaped glassware.
- Stainless steel construction
- Chromed-brass band and chain
- Holds a variety of odd-shaped glassware
Constructed of stainless steel with strong, chromed-brass band and chain. Extension arm is 152 mm (6") long.