KIMBLE® Distillation Column, Vigreux, Microscale, DWK Life Sciences
Catalog # KT746150-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Microscale vigreux distillation column
- Height:75 mm
- Cat. no.:KT746150-0000
- Thread Size:20-400
- Supplier No.:746150-0000
Specifications
About this item
Column features a [ST] 14/10 joint and indentations over the entire length to optimize vapor-liquid contact.