Self Laminating Labels
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Caresfield self-laminating direct-print laser wristbands allow printing wristbands and labels on one sheet with your laser printer.
- Laser printer required
- Produce high-quality text and graphics
- Often used for admitting purpose in healthcare facilities
- Self-laminating (protects barcodes and printed text)
- Tamper-evident, adhesive closure
- Not made with natural rubber latex
Specifications
- Description:Self laminating labels for wristband, sheet, adhesive closure
- Height:1"
- Length:8.25"
- Width:0.75"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)