Order Entry
Canada
ContactUsLinkComponent
VWR UniTranz-RT® Transport System
VWR UniTranz-RT® Transport System
Catalog # 77200-798
Supplier:  VWR International
CAS Number:  
undefined
VWR UniTranz-RT® Transport System
Catalog # 77200-798
Supplier:  VWR International
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Description:
    Universal transport media
  • Size:
    3 ml
  • Cat. no.:
    77200-798

Specifications

About this item

The UniTranz-RT® universal transport is ideal for the collection and preservation of viruses including, chlamydia, mycoplasma, and urea plasma. It is also suitable for use with antigen detection, PCR, and rapid tests when validated by test manufacturers and/or laboratory.

  • Premium medical grade plastic components improve product shelf life
  • PurFlock Ultra swab: 25-3317-U
  • Shelf life: 18 months
  • For In Vitro diagnostic use
  • Made in USA

All kits include vial, and individually wrapped sterile swabs.