Raspberry Pi 4 Computers
About this item
Ground-breaking increases in speed and performance compared to the Raspberry Pi 3 Model B+.
- Choose either 4 or 8 GB of RAM
- Single board computer only; power adapter, accessories, display cable, and microSD card available separately
For the end user, Raspberry Pi 4 Model B provides desktop performance comparable to entry-level x86 PC systems. This product’s key features include a high-performance 64-bit quad-core processor, dual-display support at resolutions up to 4K via a pair of micro-HDMI ports, hardware video decode at up to 4Kp60, 4 GB of RAM, dual-band 2.4/5.0 GHz wireless LAN, Bluetooth 5.0, Gigabit Ethernet, USB 3.0, and PoE capability (via a separate PoE HAT add-on). The dual-band wireless LAN and Bluetooth have modular compliance certification, allowing the board to be designed into end products with significantly reduced compliance testing, improving both cost and time to market.
4 Options Available Below
Product Details & Documents
Ground-breaking increases in speed and performance compared to the Raspberry Pi 3 Model B+.
- Choose either 4 or 8 GB of RAM
- Single board computer only; power adapter, accessories, display cable, and microSD card available separately
For the end user, Raspberry Pi 4 Model B provides desktop performance comparable to entry-level x86 PC systems. This product’s key features include a high-performance 64-bit quad-core processor, dual-display support at resolutions up to 4K via a pair of micro-HDMI ports, hardware video decode at up to 4Kp60, 4 GB of RAM, dual-band 2.4/5.0 GHz wireless LAN, Bluetooth 5.0, Gigabit Ethernet, USB 3.0, and PoE capability (via a separate PoE HAT add-on). The dual-band wireless LAN and Bluetooth have modular compliance certification, allowing the board to be designed into end products with significantly reduced compliance testing, improving both cost and time to market.
Documents
TDS Raspberry Pi 4
Datasheet
Product Family Options
Product Information
- AvailabilityPrice
- 470354-010Raspberry Pi 4 SBC, 4 GB RAMSupplier Number: 02AH3164
- 470354-022Raspberry Pi 4 SBC, 8 GB RAMSupplier Number: 64AH2041
- 470354-020MicroSD card, 16 GB, with Raspberry Pi OSSupplier Number: 29AK8617
- 470354-024USB-C power adapterSupplier Number: 03AH7034