Orders LinkContactUsLinkComponent
Raspberry Pi 4 Starter Kits
Supplier:   NEWARK ELECTRONICS
Raspberry Pi 4 Starter Kits
Supplier:  NEWARK ELECTRONICS
Certificates
Raspberry Pi 4 Starter Kits
Raspberry Pi 4 Starter Kits
2 Options Available Below
Product Details & Documents
Complete Raspberry Pi kit contains everything needed to get started in single board computing.
  • Perfect for makerspaces and Fab labs, as well as computer science classrooms
  • Includes a 16 GB microSD card loaded with Raspberry Pi OS for easy start up
  • Included 3 A power adapter provides enough power for advanced applications
  • Includes 3' micro HDMI to HDMI cable for simple connections to an existing display
Introducing the Raspberry Pi 4 Model B, with 4 GB RAM. At its foundation, this R-Pi offers increased features and capabilities, while maintaining full backwards compatibility with the earlier Raspberry Pi 3 Model B+. This product’s key features include a high-performance 64-bit quad-core processor, dual-display support at resolutions up to 4K via a pair of micro-HDMI ports, hardware video decode at up to 4Kp60, 4 GB of RAM, dual-band 2.4/5.0 GHz wireless LAN, Bluetooth 5.0, Gigabit Ethernet, USB 3.0, and PoE capability (via a separate PoE HAT add-on). The dual-band wireless LAN and Bluetooth have modular compliance certification, allowing the board to be designed into end products with significantly reduced compliance testing, improving both cost and time to market.
 : Kit includes: Raspberry Pi 4 model B, with 4 GB RAM, 5.1 V, 3 A power supply, 16 GB Micro SD card, preloaded with NOOBS operating system, 1M Micro HDMI to HDMI cable, two-piece heat sink kit, molded enclosure (white), getting started booklet.
Product Family Options
Product Information
  • Availability
    Price
  • 470354-000
    Raspberry Pi 4 starter kit, white, 4 GB RAM
    Raspberry Pi 4 starter kit, white, 4 GB RAM
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $175.05
    $175.05
    Kit of 1
       $175.05
    Supplier Number: 80AH9359
  • 470354-012
    Raspberry Pi 4 starter kit, black, 4 GB RAM
    Raspberry Pi 4 starter kit, black, 4 GB RAM
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $168.10
    $168.10
    Kit of 1
       $168.10
    Supplier Number: 80AH9362