Vee Gee Filtering Flasks
About this item
Filtering flasks are constructed of heavy wall borosilicate glass in order to withstand high vacuum applications.
- Made of premium borosilicate glass
- Heavy-duty rims to reduce chipping
- Permanent white graduations
- Large marking square for labeling
- Port for tubulation has an outside diameter of 10 mm
- Accepts a No. 8 rubber stopper (Not included)
Used in conjunction with vacuum filter systems, these filtering flasks feature heavy-duty rims to reduce chipping. The flasks also feature permanent white graduations and a large marking square for labeling. All flasks are sold individually.
4 Options Available Below
Product Details & Documents
Filtering flasks are constructed of heavy wall borosilicate glass in order to withstand high vacuum applications.
- Made of premium borosilicate glass
- Heavy-duty rims to reduce chipping
- Permanent white graduations
- Large marking square for labeling
- Port for tubulation has an outside diameter of 10 mm
- Accepts a No. 8 rubber stopper (Not included)
Used in conjunction with vacuum filter systems, these filtering flasks feature heavy-duty rims to reduce chipping. The flasks also feature permanent white graduations and a large marking square for labeling. All flasks are sold individually.
Product Family Options
Product Information
- CapacityMaterialO.D.×HAvailabilityPrice
- 300 mlBorosilicate glass3.4×5.5"Each$51.59Pack of 12$619.13Supplier Number: 1780T-300
- 500 mlBorosilicate glass4.1×6.7"Each$59.37Pack of 12$712.41Supplier Number: 1780T-500
- 1000 mlBorosilicate glass5.2×8.7"Each$79.89Pack of 8$639.14Supplier Number: 1780T-1000
- 2000 mlBorosilicate glass6.5×10.6"Each$123.91Pack of 4$495.66Supplier Number: 1780T-2000