KIMBLE® AIRLESS-WARE® Manifold, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Used to connect inert gas and vacuum sources
The 4 mm (0.157") PTFE vacuum valves provide grease-free fine control. Sample contacts only PTFE and borosilicate glass. Distance between valves is 100 mm (3.94"). Beams have a 12.7 mm (0.5") outer diameter (O.D.), delivery ports have a 9 mm (0.35") O.D.
Customers Who Bought This Also Bought