ContactUsLinkComponent
Accessories for Synthware Vacuum/Inert Gas Adapters, Kemtech America
Accessories for Synthware Vacuum/Inert Gas Adapters, Kemtech America
Catalog # 60005-132
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Accessories for Synthware Vacuum/Inert Gas Adapters, Kemtech America
Catalog # 60005-132
Supplier:  Kemtech America
Supplier Number:  S201530
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • ST Joint No.:
    PTFE T-bore stopcock plug, 2 mm
  • Cat. no.:
    60005-132
  • Supplier No.:
    S201530
Specifications
About this item
Kemtech replacement t-bore stopcock.
  • Borosilicate 3.3
  • T-bore
  • 1:5 taper
T-bore, Teflon®, stopcock is a direct replacement for a number of Kemtech products.