SP Bel-Art Aquet® Detergent for Glassware and Plastics, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Safe for glassware, plastics, and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware.
- Convenient, pre-portioned pouches make one gallon of 1% solution
- 3.8 L (1 gal.) bottle comes with 473 ml (16 oz.) working solution dispenser bottle
- 20 L (5 gal.) Cubitainer™ is available for frequent users
- Not for use in automatic dishwashers
It rinses quickly and dries without wiping.
Related Information
Certificates
Search by Lot NumberFocus Safety Volume 1 Issue 1 VWR® Focus Production Volume 3
Product Literature
ghs_F17094