Liquid Scintillation Vials, PET, with Screw Cap, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
PET (polyethylene terephthalate) plastic vials offer the clarity of glass and the waste disposal advantages of plastic
- Vials available as with caps attached or unattached
Vials are virtually unbreakable, with minimal background counts and low permeability to solvents.
Ordering information: Vials with attached caps are packaged in trays of 100 vials. Vials with unattached caps are packaged in bags.
Related Information
Certificates
Search by Lot NumberDWK Life Sciences Return to Research Promo 2021
Promotion Material
Wheaton Vial GUide
Product Literature
Customers Who Bought This Also Bought