Orders LinkContactUsLinkComponent
E-C Sample Vials, WHEATON®, DWK Life Sciences
E-C Sample Vials, WHEATON®, DWK Life Sciences
E-C Sample Vials, WHEATON®, DWK Life Sciences
Certificates
E-C Sample Vials, WHEATON®, DWK Life Sciences
E-C Sample Vials, WHEATON®, DWK Life Sciences
2 Options Available Below
Product Details & Documents
Organic series clear glass vials are made from Wheaton 33® low extractable borosilicate glass.
Vials provide an inert and chemically-resistant environment for harsh, sensitive, and reactive samples. Vials accommodate size 24-400 screw caps (not included).
Certifications: Vials meet ASTM Type I Class A and USP Type I standards.
Ordering information: Packed in two shrink-wrapped packs of 100 vials.
Product Family Options
Product Information
  • Capacity
    Diameter ×H
    Cap included
    Availability
    Price
  • 20.00 ml
    28×57 mm
    No
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $381.29
    $381.29
    New Customer Promotional Price
    Case of 200
       $381.29
    Supplier Number: W224809
  • 40.00 ml
    28×95 mm
    No
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $389.30
    $389.30
    New Customer Promotional Price
    Case of 200
       $389.30
    Supplier Number: W224810