Orders LinkContactUsLinkComponent
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
Supplier:   Brandtech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
Supplier:  Brandtech
Certificates
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
VITLAB® Sample Vials, PP, with Push-On Caps, BrandTech
4 Options Available Below
Product Details & Documents
General-purpose sample vials for a variety of lab applications.
  • Polypropylene
  • LDPE push-on 'snap' caps
  • Vials are autoclavable, caps are not
Translucent polypropylene vials include low-density polyethylene push-on caps. Vials and caps are suitable for microwave use.
Product Family Options
Product Information
  • Capacity
    Diameter ×H
    Cap included
    Availability
    Price
  • 5.00 ml
    20×25 mm
    Yes
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $184.96
    $184.96
    New Customer Promotional Price
    Pack of 25
       $184.96
    Supplier Number: 68594
  • 18.00 ml
    22×57 mm
    Yes
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $255.49
    $255.49
    New Customer Promotional Price
    Pack of 25
       $255.49
    Supplier Number: 68894
  • 50.00 ml
    30×97 mm
    Yes
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $119.18
    $119.18
    New Customer Promotional Price
    Pack of 10
       $119.18
    Supplier Number: 69194
  • 160.00 ml
    50×110 mm
    Yes
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $140.18
    $140.18
    New Customer Promotional Price
    Pack of 10
       $140.18
    Supplier Number: 69294