Orders LinkContactUsLinkComponent
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Supplier:   Qorpak
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Supplier:  Qorpak
Certificates
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
38 Options Available Below
Product Details & Documents
Amber screw thread sample vials offer protection from harmful light and UV rays for light sensitive products.
These general purpose glass vials are made of Type I borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.
Ordering information: Vials 36319-407 and 66175-540 are supplied pre-cleaned for volatiles. Vial 11311-522 is supplied ultra cleaned. Vials 93000-912, 93000-922, 93000-906, and 93000-928 are supplied vacuumed and ionized.
Product Family Options
Vials without Caps
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 11311-222
    Vials only
    Vials only
    1.25 ml
    14.75×22 mm
    Amber
    No
    13-425
    Discontinued
    Supplier Number: -
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    11311-222
    Small Business Enterprise
  • 11311-224
    Vials only
    Vials only
    1.85 ml
    14.75×26 mm
    Amber
    No
    13-425
    Discontinued
    Supplier Number: -
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    11311-224
    Small Business Enterprise
  • 89237-402
    Vials only
    Vials only
    4.00 ml
    15×45 mm
    Amber
    No
    13-425
    Log in for availability
    Log in for availability
    Pack of 144
    Pack of 144 - $2,080.38
    $2,080.38
    New Customer Promotional Price
    Pack of 144
       $2,080.38
    Supplier Number: GLA-05558
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89237-402
    Small Business Enterprise
  • 89204-962
    Vials only
    Vials only
    7.50 ml
    17×60 mm
    Amber
    No
    15-425
    Log in for availability
    Log in for availability
    Case of 1,584
    Case of 1,584 - $2,037.46
    $2,037.46
    New Customer Promotional Price
    Case of 1,584
       $2,037.46
    Supplier Number: GLA-00799
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89204-962
    Small Business Enterprise
  • 89422-978
    Vials only
    Vials only
    20.00 ml
    27.75×57 mm
    Amber
    No
    24-400
    Log in for availability
    Log in for availability
    Case of 576
    Case of 576 - $622.90
    $622.90
    New Customer Promotional Price
    Case of 576
       $622.90
    Supplier Number: GLA-00801
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89422-978
    Small Business Enterprise
  • 93001-766
    Vials only
    Vials only
    40.00 ml
    27×95 mm
    Amber
    No
    24-400
    Log in for availability
    Log in for availability
    Case of 72
    Case of 72 - $141.62
    $141.62
    New Customer Promotional Price
    Case of 72
       $141.62
    Supplier Number: GLA-00802
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    93001-766
    Small Business Enterprise
Amber Borosilicate Screw Thread Vials with Black Phenolic PolyCone Lined Cap
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 16087-080
    Vials with closures
    Vials with closures
    20.00 ml
    27.75×57 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $864.40
    $864.40
    New Customer Promotional Price
    Case of 144
       $864.40
    Supplier Number: GLC-01061
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    16087-080
    Product available on GSA Advantage®
    Small Business Enterprise
  • 16087-100
    Vials with closures
    Vials with closures
    40.00 ml
    27.50×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $1,043.27
    $1,043.27
    New Customer Promotional Price
    Case of 144
       $1,043.27
    Supplier Number: GLC-04892
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    16087-100
    Small Business Enterprise
Amber Borosilicate Screw Thread Vials with Black Phenolic Rubber Lined Cap
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 16087-082
    Vials with closures
    Vials with closures
    20.00 ml
    27.75×57 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $1,086.68
    $1,086.68
    New Customer Promotional Price
    Case of 144
       $1,086.68
    Supplier Number: GLC-04918
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    16087-082
    Small Business Enterprise
  • 16087-102
    Vials with closures
    Vials with closures
    40.00 ml
    27.50×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $1,154.92
    $1,154.92
    New Customer Promotional Price
    Case of 144
       $1,154.92
    Supplier Number: GLC-05181
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    16087-102
    Small Business Enterprise
Amber Borosilicate Screw Thread Vials with Green PP Hole Cap & PTFE/Silicone Septa
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 76487-228
    Cleaned and certified for volatiles
    Cleaned and certified for volatiles
    20.00 ml
    27.25×57.5 mm
    Amber
    Yes
    24-414
    Log in for availability
    Log in for availability
    Case of 80
    Case of 80 - $255.01
    $255.01
    New Customer Promotional Price
    Case of 80
       $255.01
    Supplier Number: GLC-01057
    Specifications:
    More
    Specifications
    Cat. No.
    76487-228
  • 16054-216
    Vials with closures
    Vials with closures
    40.00 ml
    27.50×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $816.74
    $816.74
    New Customer Promotional Price
    Case of 144
       $816.74
    Supplier Number: GLC-01069
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    16054-216
    Product available on GSA Advantage®
    Small Business Enterprise
  • 76076-732
    Vials with closures, cleaned for volatiles
    Vials with closures, cleaned for volatiles
    20.00 ml
    27.75×57 mm
    Amber
    Yes
    24-400
    Discontinued
    Supplier Number: GLC-01059
    Specifications:
    More
    Specifications
    Cat. No.
    76076-732
  • 36319-408
    Vials with closures
    Vials with closures
    20.00 ml
    27.75×57 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $824.90
    $824.90
    New Customer Promotional Price
    Case of 144
       $824.90
    Supplier Number: GLC-01058
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    36319-408
    Product available on GSA Advantage®
    Small Business Enterprise
  • 66175-518
    Vials with closures
    Vials with closures
    40.00 ml
    27×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $902.75
    $902.75
    New Customer Promotional Price
    Case of 144
       $902.75
    Supplier Number: GLC-01075
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    66175-518
    Product available on GSA Advantage®
    Small Business Enterprise
  • 66175-540
    Vials with closures, cleaned for volatiles
    Vials with closures, cleaned for volatiles
    40.00 ml
    27.50×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $1,064.60
    $1,064.60
    New Customer Promotional Price
    Case of 144
       $1,064.60
    Supplier Number: GLC-01076
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    66175-540
    Product available on GSA Advantage®
    Small Business Enterprise
  • 76487-230
    Cleaned and certified for volatiles
    Cleaned and certified for volatiles
    40.00 ml
    27.25×95 mm
    Amber
    Yes
    24-414
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $535.07
    $535.07
    New Customer Promotional Price
    Case of 144
       $535.07
    Supplier Number: GLC-01071
    Specifications:
    More
    Specifications
    Cat. No.
    76487-230
  • 76487-232
    Cleaned and certified for volatiles
    Cleaned and certified for volatiles
    60.00 ml
    27.25×140 mm
    Amber
    Yes
    24-414
    Log in for availability
    Log in for availability
    Case of 72
    Case of 72 - $471.81
    $471.81
    New Customer Promotional Price
    Case of 72
       $471.81
    Supplier Number: GLC-07869
    Specifications:
    More
    Specifications
    Cat. No.
    76487-232
Amber Borosilicate Screw Thread Vials with Green Thermoset F217 & PTFE Lined Cap
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 10798-846
    Vials with Closures (Unattached)
    Vials with Closures (Unattached)
    1.25 ml
    14.75×22 mm
    Amber
    Yes
    13-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $239.93
    $239.93
    New Customer Promotional Price
    Case of 144
       $239.93
    Supplier Number: GLC-08767
    Specifications:
    More
    Specifications
    Cat. No.
    10798-846
  • 35100-106
    Vials with closures
    Vials with closures
    1.85 ml
    12×35 mm
    Amber
    Yes
    8-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $671.46
    $671.46
    New Customer Promotional Price
    Case of 144
       $671.46
    Supplier Number: GLC-06786
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    35100-106
    Small Business Enterprise
  • 93000-906
    Vials with closures, vacuum and ionized
    Vials with closures, vacuum and ionized
    1.85 ml
    12×35 mm
    Amber
    Yes
    8-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $483.42
    $483.42
    New Customer Promotional Price
    Case of 144
       $483.42
    Supplier Number: GLC-07015
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    93000-906
    Small Business Enterprise
  • 10798-848
    Vials with Closures (Unattached)
    Vials with Closures (Unattached)
    1.85 ml
    14.75×26 mm
    Amber
    Yes
    13-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $219.26
    $219.26
    New Customer Promotional Price
    Case of 144
       $219.26
    Supplier Number: GLC-08768
    Specifications:
    More
    Specifications
    Cat. No.
    10798-848
  • 35100-100
    Vials with closures
    Vials with closures
    15.00 ml
    21×70 mm
    Amber
    Yes
    18-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $483.98
    $483.98
    New Customer Promotional Price
    Case of 144
       $483.98
    Supplier Number: GLC-06789
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    35100-100
    Small Business Enterprise
  • 35100-104
    Vials with closures
    Vials with closures
    4.00 ml
    15×45 mm
    Amber
    Yes
    13-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $679.98
    $679.98
    New Customer Promotional Price
    Case of 144
       $679.98
    Supplier Number: GLC-06787
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    35100-104
    Small Business Enterprise
  • 93000-922
    Vials with closures, vacuum and ionized
    Vials with closures, vacuum and ionized
    4.00 ml
    15×45 mm
    Amber
    Yes
    13-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $423.62
    $423.62
    New Customer Promotional Price
    Case of 144
       $423.62
    Supplier Number: GLC-06029
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    93000-922
    Small Business Enterprise
  • 93000-928
    Vials with closures, vacuum and ionized
    Vials with closures, vacuum and ionized
    15.00 ml
    21×70 mm
    Amber
    Yes
    18-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $911.53
    $911.53
    New Customer Promotional Price
    Case of 144
       $911.53
    Supplier Number: GLC-06031
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    93000-928
    Small Business Enterprise
  • 76487-246
    Cleaned for volatiles
    Cleaned for volatiles
    4.00 ml
    15×45 mm
    Amber
    Yes
    13-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $387.78
    $387.78
    New Customer Promotional Price
    Case of 144
       $387.78
    Supplier Number: GLC-07294
    Specifications:
    More
    Specifications
    Cat. No.
    76487-246
  • 35100-102
    Vials with closures
    Vials with closures
    7.50 ml
    17×60 mm
    Amber
    Yes
    15-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $641.75
    $641.75
    New Customer Promotional Price
    Case of 144
       $641.75
    Supplier Number: GLC-06788
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    35100-102
    Small Business Enterprise
  • 93000-912
    Vials with closures, vacuum and ionized
    Vials with closures, vacuum and ionized
    7.50 ml
    17×60 mm
    Amber
    Yes
    15-425
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $460.69
    $460.69
    New Customer Promotional Price
    Case of 144
       $460.69
    Supplier Number: GLC-06030
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    93000-912
    Small Business Enterprise
  • 76487-248
    Cleaned for volatiles
    Cleaned for volatiles
    20.00 ml
    27.25×57.5 mm
    Amber
    Yes
    24-414
    Log in for availability
    Log in for availability
    Case of 72
    Case of 72 - $292.88
    $292.88
    Case of 72
       $292.88
    Supplier Number: GLC-12579
    Specifications:
    More
    Specifications
    Cat. No.
    76487-248
  • 76487-358
    Vials with closures, vacuum and ionized
    Vials with closures, vacuum and ionized
    20.00 ml
    27.25×57.5 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $380.18
    $380.18
    New Customer Promotional Price
    Case of 144
       $380.18
    Supplier Number: GLC-06001
    Specifications:
    More
    Specifications
    Cat. No.
    76487-358
  • 11311-570
    Vials with closures
    Vials with closures
    20.00 ml
    27.75×57 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $566.29
    $566.29
    New Customer Promotional Price
    Case of 144
       $566.29
    Supplier Number: GLC-01056
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    11311-570
    Product available on GSA Advantage®
    Small Business Enterprise
  • 11311-522
    Vials with closures, ultra cleaned
    Vials with closures, ultra cleaned
    40.00 ml
    27.50×95 mm
    Amber
    Yes
    24-400
    Discontinued
    Supplier Number: -
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    11311-522
    Small Business Enterprise
  • 93001-596
    Vials with closures
    Vials with closures
    40.00 ml
    27×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $610.23
    $610.23
    New Customer Promotional Price
    Case of 144
       $610.23
    Supplier Number: GLC-01065
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    93001-596
    Product available on GSA Advantage®
    Small Business Enterprise
Amber Borosilicate Screw Thread Vials with PP Cap & PTFE Disc
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 16054-210
    Vials with closures
    Vials with closures
    20.00 ml
    27.75×57 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $588.27
    $588.27
    New Customer Promotional Price
    Case of 144
       $588.27
    Supplier Number: GLC-01053
    Specifications:
    Product available on GSA Advantage®
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    16054-210
    Product available on GSA Advantage®
    Small Business Enterprise
  • 76487-224
    Cleaned and certified for volatiles
    Cleaned and certified for volatiles
    20.00 ml
    27.25×57.5 mm
    Amber
    Yes
    24-414
    Log in for availability
    Log in for availability
    Case of 80
    Case of 80 - $266.63
    $266.63
    New Customer Promotional Price
    Case of 80
       $266.63
    Supplier Number: GLC-05646
    Specifications:
    More
    Specifications
    Cat. No.
    76487-224
  • 93001-538
    Vials with closures
    Vials with closures
    40.00 ml
    27×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $582.46
    $582.46
    New Customer Promotional Price
    Case of 144
       $582.46
    Supplier Number: GLC-05180
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    93001-538
    Small Business Enterprise
  • 76487-226
    Cleaned and certified for volatiles
    Cleaned and certified for volatiles
    40.00 ml
    27.25×95 mm
    Amber
    Yes
    24-414
    Log in for availability
    Log in for availability
    Case of 72
    Case of 72 - $225.19
    $225.19
    New Customer Promotional Price
    Case of 72
       $225.19
    Supplier Number: GLC-01063
    Specifications:
    More
    Specifications
    Cat. No.
    76487-226
  • 82027-246
    Vials with closures
    Vials with closures
    20.00 ml
    27.75×57 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $559.46
    $559.46
    New Customer Promotional Price
    Case of 144
       $559.46
    Supplier Number: GLC-04917
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    82027-246
    Small Business Enterprise
Amber Borosilicate Screw Thread Vials with White Polypropylene F217 & PTFE Lined Cap
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 76170-368
    Vials with closures
    Vials with closures
    20.00 ml
    27.25×57.5 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $316.64
    $316.64
    New Customer Promotional Price
    Case of 144
       $316.64
    Supplier Number: 242632
    Specifications:
    More
    Specifications
    Cat. No.
    76170-368
  • 76170-372
    Vials with closures
    Vials with closures
    40.00 ml
    27.75×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 144
    Case of 144 - $343.02
    $343.02
    New Customer Promotional Price
    Case of 144
       $343.02
    Supplier Number: 242635
    Specifications:
    More
    Specifications
    Cat. No.
    76170-372
Amber Borosilicate Screw Thread Vials with White PP Hole Cap & PTFE/Silicone Septa
Product Information
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • 76487-234
    Cleaned and certified for TOC <10 ppb
    Cleaned and certified for TOC <10 ppb
    40.00 ml
    27.75×95 mm
    Amber
    Yes
    24-400
    Log in for availability
    Log in for availability
    Case of 80
    Case of 80 - $519.98
    $519.98
    New Customer Promotional Price
    Case of 80
       $519.98
    Supplier Number: GLC-06161
    Specifications:
    More
    Specifications
    Cat. No.
    76487-234