ContactUsLinkComponent
Synthware Distilling Heads for Volatile Distillants, Short Path, Kemtech America
Synthware Distilling Heads for Volatile Distillants, Short Path, Kemtech America
Catalog # 60002-084
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Distilling Heads for Volatile Distillants, Short Path, Kemtech America
Catalog # 60002-084
Supplier:  Kemtech America
Supplier Number:  D541024
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Cat. no.:
    60002-084
  • Bottom Inner [ST] Joint Size:
    24/40
  • Supplier No.:
    D541024
Specifications
About this item
With Vigreux indentations
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Jacketed
  • Microscale
  • Drip tip
  • Designated 10/18 thermometer joint
Condenser is a combination of a cold coil and Leibig. Top outer joint is [ST]10/18 for use with a 50mm immersion thermometer.
Ordering information: Distillation receiver and thermometer are not supplied. For best results, use distillation receiver 60002-064. For thermometer, contact your Avantor representative.