Synthware Distilling Head for Volatile Solvents, Short Path, Kemtech America
Catalog #: 60002-080
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. no.:60002-080
- Bottom Inner [ST] Joint Size:24/40
- Supplier No.:D531024
Specifications
About this item
Condenser is a combination of a cold coil and Liebig.
- Heavy wall Borosilicate construction
- Ground glass joint
- Jacketed
- Microscale
- Drip tip
- Designated 10/18 Thermometer joint
Top outer joint is [ST]10/18 for use with a 50 mm immersion thermometer.
Ordering information: Distillation receiver and thermometer are not supplied. For best results, use distillation receiver 60002-064. For thermometer, contact your Avantor representative.
Customers Who Bought This Also Bought