Synthware Distillation Receivers without Hose Connection, Kemtech America
Supplier: Kemtech America
Certificates
About this item
With the outer and inner joints at 135° angle.
- Heavy wall borosilicate construction
- Ground glass joint
With the outer and inner joint at 135° angle, used with short-path.
Frequently Bought Together