ContactUsLinkComponent
Synthware Distillation Cow Receivers, Kemtech America
Synthware Distillation Cow Receivers, Kemtech America
Catalog # 60002-102
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Distillation Cow Receivers, Kemtech America
Catalog # 60002-102
Supplier:  Kemtech America
Supplier Number:  D161420
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Cat. no.:
    60002-102
  • ST Joint Size:
    14/20
  • Supplier No.:
    D161420
Specifications
About this item
Designed for microscale application.
  • Heavy wall borosilicate construction
  • Ground glass joint
With one female joint and four drip-tip male joints.