Synthware Distillation Apparatus, Microscale Vigreux
Supplier: Kemtech America
Certificates
About this item
With Vigreux indentations.
- Heavy wall borosilicate construction
- Ground glass joint
- Jacketed
- Microscale
- Drip tip
- Designated 10/18 thermometer joint
- Vigreux indentations
Used with 25 mm immersion thermometer and distillation receiver. Thermometer [ST] joint size: 10/18.
Ordering information: Distillation receiver and thermometer are not supplied. For distallation receiver, see 60002-060 series. For thermometer, contact your Avantor representative.
Frequently Bought Together