Large Volume Multi-Compartment Dispensers, S-Curve Technologies
Supplier: S-Curve Technologies
Certificates
About this item
Two- and three-compartment dispensers are made from clear, clean, 6mm (1/4") thick acrylic or PETG
The PETG material is safe with IPA. Front opening provides easy access to items. Ideal for large quantities of gloves, glove liners, face masks, shoe covers, and other disposables. Edges are flame-treated for smoothness. Supplied with a heavy-duty wall bracket and support bar for mounting to a wall.
Specifications
- Catalog No:
- 89208-698
- 32948-922
- 32948-924
- PPE type:
- Multi-use
- Multi-use
- Multi-use
- Material:
- PETG
- Acrylic
- Acrylic
- No. of compartments:
- 3
- 3
- 2
- Dispenser type:
- Bench top/wall-mountable
- Bench top/wall-mountable
- Bench top/wall-mountable