Order Entry
Canada
ContactUsLinkComponent
Somso® Heart with Conducting System
Somso® Heart with Conducting System
Catalog # 470140-302
Supplier:  MARCUS SOMMER
undefined
Somso® Heart with Conducting System
Catalog # 470140-302
Supplier:  MARCUS SOMMER
Supplier Number:  HS 6/1

Specifications

  • Description:
    Heart with conducting system
  • Height:
    32 cm
  • Length:
    18 cm
  • Cat. no.:
    470140-302
  • Width:
    18 cm
  • Supplier No.:
    HS 6/1

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Approximately twice natural size, in SOMSO-Plast®. Sectioned so that both ventricles and atria open to expose the valves. Large blood vessels near the heart and the heart muscles are shown. The conducting system and the excitation system of nerve tracts with the addition of the sinoauricular and atrioventricular nodes, the trunk and the atrioventricular bundle are shown. Separates into 4 parts. On a stand with green base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.