Order Entry
Canada
ContactUsLinkComponent
Somso® Flower of the Grape Vine
Somso® Flower of the Grape Vine
  470312-456
undefined
Somso® Flower of the Grape Vine
  470312-456
 :  BOS15/8

 

  • Description:
    Flower of the grape vine
  • Height:
    33 cm
  • Length:
    18 cm
  • Environmentally Preferable:
  • Cat. no.:
    470312-456
  • Width:
    18 cm

 

 

Somso® premium quality models.

  • 5 year warranty

Vitis vinifera, enlarged approx. 50 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. The corolla leaves are fused as in nature. The corolla is removable as a whole. The ovary is cut longitudinally. One part can be removed with two of the five stamens and the two ovary compartments with ovules are to be seen. Separable into 3 parts. On a stand with green base and ex-planatory note.

 : Items on this page are not stocked and may have an extended time for delivery.