Order Entry
Canada
ContactUsLinkComponent
Somso® Salvia Pratensis, Flower
Somso® Salvia Pratensis, Flower
Catalog # 470312-434
Supplier:  MARCUS SOMMER
undefined
Somso® Salvia Pratensis, Flower
Catalog # 470312-434
Supplier:  MARCUS SOMMER
Supplier Number:  BOS15/1

Specifications

  • Description:
    Meadow clary
  • Height:
    36 cm
  • Length:
    33 cm
  • Environmentally Preferable:
  • Cat. no.:
    470312-434
  • Width:
    18 cm

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Enlarged approx. 15 times, in SOMSO-Plast®. After Prof. Dr. W. Jung and Prof. Dr. W. Weber. The mechanism of the stamens can be demonstrated. In one piece. On a stand with green base and explanatory note.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.