Order Entry
Canada
ContactUsLinkComponent
Somso® Vertebral Column with Pelvis
Somso® Vertebral Column with Pelvis
Catalog # 470105-206
Supplier:  MARCUS SOMMER
undefined
Somso® Vertebral Column with Pelvis
Catalog # 470105-206
Supplier:  MARCUS SOMMER
Supplier Number:  QS 21/1

Specifications

  • Description:
    Vertebral column with pelvis
  • Height:
    83 cm
  • Length:
    19 cm
  • Cat. no.:
    470105-206
  • Width:
    28 cm
  • Supplier No.:
    QS 21/1

Specifications

About this item

Somso® premium quality models.

  • 5 year warranty

Cast from natural specimen, in SOMSO-Plast®. Mounted and flexible, showing the arteria vertebralis, the spinal cord and the spinal nerves leaving it and the connected ganglion cells. Consisting of occipital bone, cervical, thoracic and lumbar vertebrae, sacrum and coccyx and innominates. Especially suitable for the demonstration of the curvature of healthy and pathological vertebral columns.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.