Order Entry
Canada
Orders LinkContactUsLinkComponent
Somso® Rape Pod

Somso® Rape Pod

Rape pod, 18 cm, 18 cm, 51 cm
Catalog # 470312-526
Supplier:   MARCUS SOMMER
Supplier #:  BOS15/12
Somso® Rape Pod
Catalog # 470312-526
Supplier:  MARCUS SOMMER
Somso® Rape Pod

Rape pod, 18 cm, 18 cm, 51 cm

Price
$1,010.00
Log in for availability
Each
Product Details & Documents

Somso® premium quality models.

  • 5 year warranty

Brassica napus, enlarged approx. 8 times, in SOMSO-Plast®. After Prof. Dr. W. Weber. This model shows the typical architecture of a cruciferous plant pod. One of the two carpels can be separated from the placenta to which the seeds are attached. The pseudo septum with the seed containing placentas are also removable. Separable into 4 parts. On a green base with explanatory note.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.

Specifications
  • Description:
    Rape pod
  • Height:
    51 cm
  • Length:
    18 cm
  • Width:
    18 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.